Order Entry
Northern Ireland
ContactUsLinkComponent
2-Pyridylzinc bromide 0.5 M in THF, AcroSeal®
Flammable
Corrosive
Irritant
Health
2-Pyridylzinc bromide 0.5 M in THF, AcroSeal®
Catalog # 433630500.
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
2-Pyridylzinc bromide 0.5 M in THF, AcroSeal®
Catalog # 433630500.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Dark green to brown to black Solution
  • Titration Argentometric:
    0.45 to 0.65 M
  • Water:
    ≤0.005 %
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.001 %

Specification Test Results

About this item

 :
 :223.378 g/mol
Density:0,974 g/cm³
Flash Pt:–17 °C
Storage Temperature:Refrigerator
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:218777-23-2
Formula:C₅H₄BrNZn
MDL Number:MFCD00671985