Order Entry
Northern Ireland
ContactUsLinkComponent
tert-Butylchlorodimethylsilane 50% (w/w) in toluene, AcroSeal®
Flammable
Corrosive
Irritant
Health
Environment
tert-Butylchlorodimethylsilane 50% (w/w) in toluene, AcroSeal®
Catalog # 433111000.
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Environment
tert-Butylchlorodimethylsilane 50% (w/w) in toluene, AcroSeal®
Catalog # 433111000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Assay:
    48 to 52 %
  • Melting point:
    118 to 122 °C
  • Titration with NaOH:
    ≥97.5 %

Specification Test Results

About this item

 :
 :150.72 g/mol
Boiling Pt:110…126 °C (760 mmHg)
Density:0,866 g/cm³
Flash Pt:4 °C
Storage Temperature:Ambient
UN:2920
ADR:8,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:18162-48-6
Formula:C₆H₁₅ClSi
MDL Number:MFCD00000501