


Morpholine 99.5%, AcroSeal®, purified by redistillation
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-91-8
:
Formula:C₄H₉NO
MDL Number:MFCD00005972
:87.12 g/mol
Boiling Pt:126…130 °C (760 mmHg)
Melting Pt:–5 °C
Density:0,99 g/cm³
Flash Pt:32 °C
UN:2054
ADR:8,I
:GHS06
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4530 to 1.4550 (20°C, 589 nm)
- GC:≥99.4 %
- Color scale:≤20 APHA
Specification Test Results
Related Information
Certificates
Search by Lot NumberACRO43385
SDS
AcroSeal Brochure VWR Final AO_AcroSeal_Product Lists_VWR_final
Product Literature
Frequently Bought Together