



Lithium diphenylphosphide 0.5 M in THF, AcroSeal®
Catalog # 433621000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Orange to red Liquid
- Titration with HClO4:0.40 to 0.60 M
- Water:≤0.005 %
- GC:≥99.0 %
- Residue after evaporation:≤0.001 %
Specification Test Results
About this item
:
Boiling Pt:65…67 °C (760 mmHg)
Density:0,925 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS05
:GHS07
:GHS02
:GHS08
CAS Number:65567-06-8
:4541-02-0
Formula:C₁₂H₁₀LiP
MDL Number:MFCD00046054