Order Entry
Northern Ireland
ContactUsLinkComponent
Chlorodimethyl-3,3,3-trifluoropropylsilane 95%
Corrosive
Flammable
Chlorodimethyl-3,3,3-trifluoropropylsilane 95%
Catalog # ACRO432210050
CAS Number:  
undefined
Corrosive
Flammable
Chlorodimethyl-3,3,3-trifluoropropylsilane 95%
Catalog # ACRO432210050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to straw yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3720 to 1.3780 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

About this item

 :
 :190.67 g/mol
Density:1,11 g/cm³
Flash Pt:20 °C
Storage Temperature:Ambient
UN:2986
ADR:8,II
 :GHS05
 :GHS02
CAS Number:1481-41-0
Formula:C₅H₁₀ClF₃Si
MDL Number:MFCD00053211