



3,5-Dimethylphenylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431940500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Yellow to brown Solution
- Titration Grignard:0.40 to 0.60 M
- Melting point:96 to 99 °C
- HPLC:≥98.5 %
Specification Test Results
About this item
:
:209.37 g/mol
Density:0,91 g/cm³
Flash Pt:–18 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:34696-73-6
Formula:C₈H₉BrMg
MDL Number:MFCD01311497