Order Entry
Northern Ireland
ContactUsLinkComponent
(1,3-Dioxan-2-ylethyl)magnesium bromide 0.5 M in THF, AcroSeal®
Flammable
Corrosive
Irritant
Health
(1,3-Dioxan-2-ylethyl)magnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431900500.
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
(1,3-Dioxan-2-ylethyl)magnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431900500.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Light yellow to brown Solution (May develop some turbidity or precipitate)
  • Titration Grignard:
    0.40 to 0.60 M
  • Melting point:
    64 to 70 °C
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :219.36 g/mol
Density:0,951 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:78078-50-9
Formula:C₆H₁₁BrMgO₂
MDL Number:MFCD00061518