Order Entry
Northern Ireland
ContactUsLinkComponent
[2-(1-Piperidinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Irritant
Corrosive
Flammable
Health
[2-(1-Piperidinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Catalog # 431710500.
CAS Number:  
undefined
Irritant
Corrosive
Flammable
Health
[2-(1-Piperidinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Catalog # 431710500.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Brown Solution
  • Titration Grignard:
    0.15 to 0.35 M
  • Melting point:
    88 to 92 °C
  • GC:
    ≥98.0 %

Specification Test Results

About this item

 :
 :278.476 g/mol
Density:0,922 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS07
 :GHS05
 :GHS02
 :GHS08
CAS Number:480424-81-5
Formula:C₁₂H₁₆BrMgN
MDL Number:MFCD03840894