



4-Isopropylphenylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431630500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Yellow to brown Solution
- Titration Grignard:0.40 to 0.60 M
- HPLC:≥96.0 %
- Titration with NaOH:≥96.0 %
Specification Test Results
About this item
:
:223.396 g/mol
Density:0,939 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:18620-03-6
Formula:C₉H₁₁BrMg
MDL Number:MFCD01319911