Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w), AcroSeal®
Irritant
Health
Corrosive
Flammable
Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w), AcroSeal®
Certificates
undefined
Irritant
Health
Corrosive
Flammable
Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w), AcroSeal®
Certificates

Specification Test Results

  • Appearance:
    Clear to slightly hazy colorless to amber Viscous liquid
  • Assay:
    68.0 to 72.0 %
  • Melting point:
    50 to 54 °C
  • GC:
    ≥98.5 %

Specification Test Results

Frequently Bought Together