


Methyl chloroformate 99%, AcroSeal®
Catalog # 430841000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3860 to 1.3880 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
About this item
:
:94.5 g/mol
Boiling Pt:70…72 °C (760 mmHg)
Melting Pt:–61 °C
Density:1,223 g/cm³
Flash Pt:5 °C
UN:1238
ADR:6.1,I
:GHS06
:GHS05
:GHS02
Storage Temperature:Refrigerator
CAS Number:79-22-1
Formula:C₂H₃ClO₂
MDL Number:MFCD00000639