
4-Nitro-1H-pyrazole 97%
ACRO430160050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:160 to 165 °C
- HPLC:≥96.0 %
- UV transmittance:≥99 % (c=0.01M water, 254 nm)
:
:113.0800 g/mol
:161…162 °C
:GHS07
:Ambient
:2075-46-9
:C₃H₃N₃O₂
:MFCD00159626