
2-Pyrimidinecarbonitrile 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14080-23-0
:
Formula:C₅H₃N₃
MDL Number:MFCD00160513
:105.1 g/mol
Melting Pt:37…41 °C
Flash Pt:110 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to off-white Crystalline powder or low melting solid
- Infrared spectrum:Conforms
- Melting point:35 to 42 °C
- GC:≥97.5 %
- Specific optical rotation:+96° to +107° (20 °C, 589 nm) (C=1, EtOH)
Specification Test Results
Frequently Bought Together