Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium ethoxide 21% in ethanol, AcroSeal®
Corrosive
Flammable
Sodium ethoxide 21% in ethanol, AcroSeal®
Catalog # 429301000.
CAS Number:  
undefined
Corrosive
Flammable
Sodium ethoxide 21% in ethanol, AcroSeal®
Catalog # 429301000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Solution (May darken on storage)
  • Titration with HCl:
    20.0 to 22.0 %
  • NaOH + Na2CO3 (as NaOH):
    ≤0.5 %
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :68.05 g/mol
Boiling Pt:91 °C
Density:0,868 g/cm³
Flash Pt:25 °C
UN:2924
ADR:3,II
 :GHS05
 :GHS02
Storage Temperature:Ambient
CAS Number:141-52-6
Formula:C₂H₅NaO
MDL Number:MFCD00012417