


Triethyloxonium tetrafluoroborate 1M in methylene chloride, AcroSeal®
Catalog # 429161000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear to slightly hazy colorless to yellow to pink to brown Solution (May develop some turbidity or precipitate)
- Titration with NaOH:0.90 to 1.10 M
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥97.0 %
Specification Test Results
About this item
:
:189.99 g/mol
Density:1,328 g/cm³
Storage Temperature:Freezer
UN:2922
ADR:8,II
:GHS05
:GHS07
:GHS08
CAS Number:368-39-8
Formula:C₆H₁₅BF₄O
MDL Number:MFCD00044423