Order Entry
Northern Ireland
ContactUsLinkComponent
2-(4-Chlorosulphonylphenyl)ethyltrimethoxysilane 50% in dichloromethane, AcroSeal®
Corrosive
Irritant
Health
2-(4-Chlorosulphonylphenyl)ethyltrimethoxysilane 50% in dichloromethane, AcroSeal®
Catalog # 428991000.
CAS Number:  
undefined
Corrosive
Irritant
Health
2-(4-Chlorosulphonylphenyl)ethyltrimethoxysilane 50% in dichloromethane, AcroSeal®
Catalog # 428991000.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to light brown Solution
  • Infrared spectrum:
    Conforms
  • Assay:
    45 to 55 % solids

Specification Test Results

About this item

 :
 :324.847 g/mol
Boiling Pt:>40 °C (760 mmHg)
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :GHS05
 :GHS07
 :GHS08
CAS Number:126519-89-9
Formula:C₁₁H₁₇ClO₅SSi
MDL Number:MFCD00054774