2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane 97%
:331465-71-5
:
:C₁₈H₃₉N₄P
:MFCD03840347
:110…118 °C (,5 mmHg)
:0,964 g/cm³
:68 °C
:342.51 g/mol
:GHS07
:Ambient
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Elemental Analysis:60.60 to 65.65 % C
Frequently Bought Together