
2-Amino-4'-methoxyacetophenone hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3883-94-1
:
Formula:C₉H₁₂ClNO₂
MDL Number:MFCD00193078
:201.65 g/mol
Melting Pt:190…193 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to tan Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together