
3-Bromo-1-(trimethylsilyl)-1-propyne 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38002-45-8
:
Formula:C₆H₁₁BrSi
MDL Number:MFCD00134460
:191.14 g/mol
Boiling Pt:44…45 °C (2 mmHg)
Density:1,351 g/cm³
Flash Pt:62 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4820 to 1.4840 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together