
4-Methoxy-o-phenylenediamine dihydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59548-39-9
:
Formula:C₇H₁₀N₂O·2HCl
MDL Number:MFCD00052003
:211.09 g/mol
Melting Pt:206…209 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Violet Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 to ≤102.5 %
Specification Test Results
Frequently Bought Together