Order Entry
Northern Ireland
ContactUsLinkComponent
 
Polyvinyl alcohol, M.W. ∼ 20000-30000 88%, hydrolysed
 
undefined
Polyvinyl alcohol, M.W. ∼ 20000-30000 88%, hydrolysed

 

  • Appearance:
    Off-white to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Viscosity:
    3.5 to 4.5 cP (4 % in water at 20°C) (Brookfield)
  • Degree of hydrolysis:
    87.0 to 89.0 %
  • Ash:
    ≤0.9 % (Na2O)
  • pH:
    4.5 to 6.5 (4 % in water)
  • Volatile matter:
    ≤5.0 % (105°C, 3 h)

 

 

Frequently Bought Together