

3-Picolyl bromide hydrobromide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4916-55-6
:
Formula:C₆H₇Br₂N
MDL Number:MFCD01321314
:252.94 g/mol
Melting Pt:150…155 °C
UN:2923
ADR:8,II
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:148 to 154 °C
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together