tert-Butyllithium 1.9 M in pentane, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:594-19-4
:
Formula:C₄H₉Li
MDL Number:MFCD00008795
:64.06 g/mol
Boiling Pt:36…38 °C (760 mmHg)
Density:0,69 g/cm³
Flash Pt:–56 °C
UN:3394
ADR:4.2,
:GHS07
:GHS08
:GHS05
:GHS09
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
- Titration Active base:17.0 to 19.0 %
Specification Test Results
Related Information
Frequently Bought Together