
1,2,3-Heptanetriol 95% , high melting isomer, mixture of diastereoisomers
Catalog # ACRO395450010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:75 to 81 °C
- GC:≥94 %
Specification Test Results
About this item
:
:148.202 g/mol
Melting Pt:92…97 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:103404-57-5
Formula:C₇H₁₆O₃
MDL Number:MFCD00010517