5-Bromopicolinic acid 97%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid
- CAS Number:30766-11-1
- Formula:C₆H₄BrNO₂
- MDL Number:MFCD00234149
- Molecular Weight:202.01 g/mol
- Melting Pt:177…178 °C
- Storage Temperature:Refrigerator
3 Options Available Below
Product Details & Documents
Synonyms: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid
- CAS Number:30766-11-1
- Formula:C₆H₄BrNO₂
- MDL Number:MFCD00234149
- Molecular Weight:202.01 g/mol
- Melting Pt:177…178 °C
- Storage Temperature:Refrigerator
Documents
ACRO39399
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



