
3-Bromo-4-methylpyridine 97%
:3430-22-6
:
:C₆H₆BrN
:MFCD00082592
:172.02 g/mol
:76 °C (10 mmHg)
:1,549 g/cm³
:79 °C
:GHS07
:Ambient
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5600 to 1.5640 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together