Order Entry
Northern Ireland
ContactUsLinkComponent
Lithium phenoxide 0.6 M in THF, AcroSeal®
Corrosive
Flammable
Irritant
Health
Lithium phenoxide 0.6 M in THF, AcroSeal®
Catalog # 388611000.
CAS Number:  
undefined
Corrosive
Flammable
Irritant
Health
Lithium phenoxide 0.6 M in THF, AcroSeal®
Catalog # 388611000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to orange Solution
  • Titration with HClO4:
    0.55 to 0.65 M

Specification Test Results

About this item

 :
Boiling Pt:67 °C (760 mmHg)
Density:0,918 g/cm³
Flash Pt:–19 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS05
 :GHS02
 :GHS07
 :GHS08
CAS Number:555-24-8
Formula:C₆H₅LiO
MDL Number:MFCD00013473