Order Entry
Northern Ireland
ContactUsLinkComponent
Lithium isopropoxide 1 M in hexanes, AcroSeal®
Irritant
Health
Corrosive
Environment
Flammable
Lithium isopropoxide 1 M in hexanes, AcroSeal®
Catalog # 380201000.
CAS Number:  
undefined
Irritant
Health
Corrosive
Environment
Flammable
Lithium isopropoxide 1 M in hexanes, AcroSeal®
Catalog # 380201000.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow to orange Solution
  • Titration with HClO4:
    0.95 to 1.15 M

Specification Test Results

About this item

 :
 :66.028 g/mol
Density:0,69 g/cm³
Flash Pt:–27 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS07
 :GHS08
 :GHS05
 :GHS09
 :GHS02
CAS Number:2388-10-5
Formula:C₃H₇LiO
MDL Number:MFCD00048286