2(5H)-Furanone 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:497-23-4
:
Formula:C₄H₄O₂
MDL Number:MFCD00005376
Boiling Pt:212…214 °C (760 mmHg)
Melting Pt:3…4 °C
Density:1,19 g/cm³
Flash Pt:101 °C
:84.07 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow to amber Liquid
- Infrared spectrum:Conforms
- GC:≥95.0 %
Specification Test Results
Frequently Bought Together