Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
(Triphenylphosphoranylidene)acetonitrile 96%
  ACRO371570050
 :  
undefined
 
(Triphenylphosphoranylidene)acetonitrile 96%
  ACRO371570050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Off-white to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    189 to 198 °C
  • Additional info:
    This product unavoidably and progressively degrades
  • HPLC:
    ≥95 %

 

 

 :
 :189…198 °C
 :Freezer
 :6.1,III
 :301.33 g/mol
 :GHS07
 :16640-68-9
 :C₂₀H₁₆NP
 :MFCD00567633