Order Entry
Northern Ireland
ContactUsLinkComponent
2,3-O-Isopropylidene-D-glyceraldehyde 97%
2,3-O-Isopropylidene-D-glyceraldehyde 97%
Catalog # ACRO369260010
CAS Number:  
undefined
2,3-O-Isopropylidene-D-glyceraldehyde 97%
Catalog # ACRO369260010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Clear to hazy colorless to yellow Liquid or solid
  • Infrared spectrum:
    Conforms
  • Additional info:
    Contains various amounts of polymer
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    +39° to +69° (20°C, 589 nm) (C=2, Chloroform)

Specification Test Results

About this item

 :
 :130.143 g/mol
Boiling Pt:139 °C (760 mmHg)
Density:1,045 g/cm³
Flash Pt:62 °C
Storage Temperature:Freezer
CAS Number:15186-48-8
Formula:C₆H₁₀O₃
MDL Number:MFCD00269682