Order Entry
Northern Ireland
ContactUsLinkComponent
tert-Butylperoxy isopropyl carbonate 75% in aromatic free mineral spirit
Flammable
Irritant
Health
Environment
tert-Butylperoxy isopropyl carbonate 75% in aromatic free mineral spirit
Catalog # ACRO349842500
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
tert-Butylperoxy isopropyl carbonate 75% in aromatic free mineral spirit
Catalog # ACRO349842500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    Clear solution
  • Titration Iodometric:
    74 to 76 %
  • Color scale:
    ≤10 APHA
  • Hydrolyzable chloride:
    ≤150 ppm Inorganic + organic
  • Impurity:
    ≤1000 ppm Hydroperoxides (as TBHP)

Specification Test Results

About this item

 :
 :176.212 g/mol
Melting Pt:–20 °C
Density:0,905 g/cm³
Flash Pt:47 °C
Storage Temperature:Refrigerator
UN:3103
ADR:5.2,
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:2372-21-6
Formula:C₈H₁₆O₄
MDL Number:MFCD00126878