Order Entry
Northern Ireland
ContactUsLinkComponent
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide 98+%
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide 98+%
Catalog # ACRO294580010
CAS Number:  
undefined
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide 98+%
Catalog # ACRO294580010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    168 to 174 °C
  • NMR:
    ≥98.0 %
  • Specific optical rotation:
    +44° to +48° (20 °C, 589 nm) (C=2, Chloroform)

Specification Test Results

About this item

 :
 :229.29 g/mol
Melting Pt:170 °C
Storage Temperature:Refrigerator
CAS Number:104322-63-6
Formula:C₁₀H₁₅NO₃S
MDL Number:MFCD00075319