
2-Hydroxypyridine-N-oxide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13161-30-3
:
Formula:C₅H₅NO₂
MDL Number:MFCD00006195
:111.10 g/mol
Melting Pt:147…152 °C
Flash Pt:100 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:147 to 152 °C
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together