



2,4,6-Trimethylphenylmagnesium bromide 1 M in THF, AcroSeal®
210731000.
:
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
- Appearance:Light yellow to brown Solution
- Titration Grignard:0.95 to 1.10 M
:
:223.396 g/mol
:1,005 g/cm³
:–17 °C
:Ambient
:2924
:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:2633-66-1
:C₉H₁₁BrMg
:MFCD00009663