


Bromomethyl methyl ether 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13057-17-5
:
Formula:C₂H₅BrO
MDL Number:MFCD00000171
:124.97 g/mol
Boiling Pt:87 °C (760 mmHg)
Density:1,53 g/cm³
Flash Pt:26 °C
UN:1992
ADR:3,III
:GHS02
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4550 to 1.4570 (20°C, 589 nm)
- GC:≥88 %
Specification Test Results
Frequently Bought Together