


4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) 95%
Catalog # ACRO159342500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid or low melting solid
- Infrared spectrum:Conforms
- GC:≥94 %
- UV:(1 M in water)
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤0.001 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
About this item
:
Boiling Pt:325…327 °C (760 mmHg)
Melting Pt:36,5 °C
Flash Pt:159 °C
ADR:8,II
:210.37 g/mol
UN:3259
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:1761-71-3
Formula:C₁₃H₂₆N₂
MDL Number:MFCD00001496