


4-Butylaniline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-13-2
:
Formula:C₁₀H₁₅N
MDL Number:MFCD00007925
:149.24 g/mol
Boiling Pt:133…134 °C (14 mmHg)
Melting Pt:–14 °C
Density:0,945 g/cm³
Flash Pt:101 °C
UN:2735
ADR:8,II
:GHS07
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5340 to 1.5360 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together