Stigmasterol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:83-48-7
:
Formula:C₂₉H₄₈O
MDL Number:MFCD00003630
Molecular Weight:412.70 g/mol
Melting Pt:161…170 °C
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:161 to 170 °C
- Loss on drying:≤1 %
- GC:≥94 %
- Specific optical rotation:-48° to -52° (20 °C, 589 nm) (c=2, CHCl3)
Specification Test Results
Frequently Bought Together