
N-Methyl-3-pyrrolidinol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13220-33-2
:
Formula:C₅H₁₁NO
MDL Number:MFCD00003176
:101.15 g/mol
Boiling Pt:50…52 °C (1 mmHg)
Density:0,921 g/cm³
Flash Pt:71 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow to golden Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4630 to 1.4650 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together