2-Hydrazinylpyridine 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 2(1H)-Pyridinone, hydrazone, 2-hydrazinylpyridine, 2-Hydrazinopyridine
- CAS Number:4930-98-7
- Formula:C₅H₇N₃
- MDL Number:MFCD00006249
- Molecular Weight:109.13 g/mol
- Boiling Pt:185 °C (140 mmHg)
- Melting Pt:44,5…46 °C
- Flash Pt:110 °C
- Storage Temperature:Refrigerator
3 Options Available Below
Product Details & Documents
Synonyms: 2(1H)-Pyridinone, hydrazone, 2-hydrazinylpyridine, 2-Hydrazinopyridine
- CAS Number:4930-98-7
- Formula:C₅H₇N₃
- MDL Number:MFCD00006249
- Molecular Weight:109.13 g/mol
- Boiling Pt:185 °C (140 mmHg)
- Melting Pt:44,5…46 °C
- Flash Pt:110 °C
- Storage Temperature:Refrigerator
Specifications for Product Family
Specification Test Results
- Appearance:White to orange-brown Crystals or crystalline mass
- Infrared spectrum:Conforms
- Melting point:43 to 47 °C
- GC:≥97.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



