Order Entry
Northern Ireland
ContactUsLinkComponent
6-Aminonicotinamide 98%
Health
6-Aminonicotinamide 98%
Certificates
undefined
Health
6-Aminonicotinamide 98%

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    White to light yellow to light grey Crystalline powder or crystals or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    245 to 248 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together