


Isophorone diisocyanate (mixture of isomers) 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4098-71-9
:
Formula:C₁₂H₁₈N₂O₂
MDL Number:MFCD00064956
:222.29 g/mol
Boiling Pt:153 °C (10 mmHg)
Flash Pt:163 °C
UN:2290
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4830 to 1.4850 (20°C, 589 nm)
- GC:≥98.0 % (sum of mixed isomers)
Specification Test Results
Frequently Bought Together