

Isovaleronitrile 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:625-28-5
:
Formula:C₅H₉N
MDL Number:MFCD00001944
:83.13 g/mol
Boiling Pt:128…130 °C (730 mmHg)
Density:0,795 g/cm³
Flash Pt:28 °C
Storage Temperature:Refrigerator
UN:1992
ADR:3,III
:GHS02
:GHS06
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3920 to 1.3940 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together