Order Entry
Northern Ireland
ContactUsLinkComponent
3,5-Dimethoxybenzyl bromide 95%
3,5-Dimethoxybenzyl bromide 95%
Catalog # ACRO427340050
CAS Number:  
undefined
3,5-Dimethoxybenzyl bromide 95%
Catalog # ACRO427340050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to brown Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    68 to 72 °C
  • HPLC:
    ≥94 %

Specification Test Results

About this item

 :
 :231.0864 g/mol
Melting Pt:69…70 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
 :GHS05
CAS Number:877-88-3
Formula:C₉H₁₁BrO₂
MDL Number:MFCD01321368