Order Entry
Northern Ireland
ContactUsLinkComponent
Ethylenediamine 99+%, AcroSeal®
Flammable
Corrosive
Toxic
Health
Ethylenediamine 99+%, AcroSeal®
Certificates
undefined
Flammable
Corrosive
Toxic
Health
Ethylenediamine 99+%, AcroSeal®

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4555 to 1.4575 (20°C, 589 nm)
  • Water:
    ≤0.5 % (Coulometric)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤30 APHA

Specification Test Results

Frequently Bought Together