Order Entry
Northern Ireland
ContactUsLinkComponent
1,4-Pentadien-3-ol 98% stabilised
Irritant
Flammable
1,4-Pentadien-3-ol 98% stabilised
Certificates
undefined
Irritant
Flammable
1,4-Pentadien-3-ol 98% stabilised

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4440 to 1.4460 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Stabilizer:
    Approx. 0.4 % Hydroquinone

Specification Test Results

Related Information

Frequently Bought Together