5-tert-Butyl-N-carbobenzoxy-L-glutamate 98+%
Catalog # ACRO426150050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:86 to 91 °C
- HPLC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
- Specific optical rotation:-13° to -15° (20 °C, 589 nm) (c=2, CH3OH)
Specification Test Results
About this item
CAS Number:3886-08-6
:
Formula:C₁₇H₂₃NO₆
MDL Number:MFCD00038274
:337.37 g/mol
Melting Pt:85 °C
Storage Temperature:Ambient