
2-Methyl-3-buten-1-ol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4516-90-9
:
Formula:C₅H₁₀O
MDL Number:MFCD00009985
:86.134 g/mol
Boiling Pt:120…121 °C (760 mmHg)
Density:0,835 g/cm³
Flash Pt:33 °C
Storage Temperature:Refrigerator
UN:1987
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4240 to 1.4300 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together