Order Entry
Northern Ireland
ContactUsLinkComponent
Trimethyltin chloride 20% (w/w) 1 M in THF, AcroSeal®
Toxic
Environment
Flammable
Irritant
Health
Trimethyltin chloride 20% (w/w) 1 M in THF, AcroSeal®
Catalog # 427851000.
CAS Number:  
undefined
Toxic
Environment
Flammable
Irritant
Health
Trimethyltin chloride 20% (w/w) 1 M in THF, AcroSeal®
Catalog # 427851000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Strength:
    0.90 to 1.10 M
  • GC:
    ≥98.0 %

Specification Test Results

About this item

 :
 :199.27 g/mol
Flash Pt:–17 °C
UN:1992
ADR:3,II
 :GHS06
 :GHS09
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:1066-45-1
Formula:C₃H₉ClSn
MDL Number:MFCD00000520